
6-(tert-Butyl)-4-chlorothieno[3,2-d]pyrimidine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:439693-52-4
:
Formula:C₁₀H₁₁ClN₂S
MDL Number:MFCD03407483
:226.726 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together