2-(1H-Pyrazol-1-yl)isonicotinonitrile
Catalog # APOSOR110591-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:170.171 g/mol
Storage Temperature:Ambient
CAS Number:956769-58-7
Formula:C₉H₆N₄
MDL Number:MFCD09373409