Methyl [5-(2-amino-1,3-thiazol-4-yl)-1-methyl-1H-pyrrol-2-yl]acetate
Catalog # APOSOR110578-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:251.305 g/mol
Storage Temperature:Ambient
CAS Number:1227954-54-2
Formula:C₁₁H₁₃N₃O₂S
MDL Number:MFCD11973598