Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butyl 4-{2-[(2-chloropyridin-3-yl)oxy]ethyl}piperazine-1-carboxylate
tert-Butyl 4-{2-[(2-chloropyridin-3-yl)oxy]ethyl}piperazine-1-carboxylate
Catalog # APOSOR110563-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 4-{2-[(2-chloropyridin-3-yl)oxy]ethyl}piperazine-1-carboxylate
Catalog # APOSOR110563-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :341.833 g/mol
Storage Temperature:Ambient
CAS Number:1227954-32-6
Formula:C₁₆H₂₄ClN₃O₃
MDL Number:MFCD11111411