
3-(Dimethylamino)-1-(2-nitrophenyl)prop-2-en-1-one
Catalog # APOSOR110438-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:220.225 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:153813-81-1
Formula:C₁₁H₁₂N₂O₃
MDL Number:MFCD00086507