
2,5-Dibromo-1H-imidazol-4-amine
Catalog # APOSOR110349-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:240.884 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1029720-13-5
Formula:C₃H₃Br₂N₃
MDL Number:MFCD10001383