Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromoimidazo[1,2-a]pyridine-2-carbohydrazide
3-Bromoimidazo[1,2-a]pyridine-2-carbohydrazide
3-Bromoimidazo[1,2-a]pyridine-2-carbohydrazide
APOSOR110330-1G
:
Apollo Scientific
:
3-Bromoimidazo[1,2-a]pyridine-2-carbohydrazide
APOSOR110330-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
255.071 g/mol
:
GHS07
:
Ambient
:
1355170-89-6
:
C₈H₇BrN₄O
:
MFCD16036201