Order Entry
Czech Republic
ContactUsLinkComponent
4-(2,4-Dichlorophenyl)-1,3-thiazol-2(3H)-one
4-(2,4-Dichlorophenyl)-1,3-thiazol-2(3H)-one
Catalog # APOSOR110317-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(2,4-Dichlorophenyl)-1,3-thiazol-2(3H)-one
Catalog # APOSOR110317-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :246.113 g/mol
Storage Temperature:Ambient
CAS Number:42489-55-4
Formula:C₉H₅Cl₂NOS
MDL Number:MFCD09746489