Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 4-hydroxy-2-methyl-1,3-thiazole-5-carboxylate 98%
Ethyl 4-hydroxy-2-methyl-1,3-thiazole-5-carboxylate 98%
Catalog # APOSOR110310-5G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 4-hydroxy-2-methyl-1,3-thiazole-5-carboxylate 98%
Catalog # APOSOR110310-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :187.216 g/mol
Storage Temperature:Ambient
CAS Number:20737-48-8
 :89776-82-9
Formula:C₇H₉NO₃S
MDL Number:MFCD08275722