Order Entry
Czech Republic
ContactUsLinkComponent
3-(Dimethylamino)-1-(2,4-dimethylphenyl)prop-2-en-1-one, tech
3-(Dimethylamino)-1-(2,4-dimethylphenyl)prop-2-en-1-one, tech
Catalog # APOSOR110206-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Dimethylamino)-1-(2,4-dimethylphenyl)prop-2-en-1-one, tech
Catalog # APOSOR110206-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :203.28 g/mol
Storage Temperature:Ambient
Formula:C₁₃H₁₇NO
MDL Number:MFCD11520130