Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Ethyl 6,8-dibromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate
Catalog # APOSOR110166-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 6,8-dibromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate
Catalog # APOSOR110166-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :362.017 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:859787-43-2
Formula:C₁₁H₁₀Br₂N₂O₂
MDL Number:MFCD22421937