(2E)-1-(4-Chlorophenyl)-3-(dimethylamino)prop-2-en-1-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:28587-05-5
Formula:C₁₁H₁₂ClNO
MDL Number:MFCD00172116
:209.68 g/mol
Storage Temperature:Ambient
Frequently Bought Together