
3-Bromoimidazo[1,2-a]pyridine-7-carbohydrazide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1427460-82-9
:
Formula:C₈H₇BrN₄O
MDL Number:MFCD21602378
:255.071 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together