
Ethyl 8-bromoimidazo[1,2-a]pyridine-6-carboxylate
Catalog # APOSOR110093-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:269.095 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:957103-97-8
Formula:C₁₀H₉BrN₂O₂
MDL Number:MFCD23159159