Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 7-bromoimidazo[1,2-a]pyridine-3-carboxylate
Irritant
Ethyl 7-bromoimidazo[1,2-a]pyridine-3-carboxylate
Catalog # APOSOR110091-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 7-bromoimidazo[1,2-a]pyridine-3-carboxylate
Catalog # APOSOR110091-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :269.095 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1134327-98-2
Formula:C₁₀H₉BrN₂O₂
MDL Number:MFCD11045695