0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
[4-(Pyrrolidin-1-ylmethyl)phenyl]methanol
[4-(Pyrrolidin-1-ylmethyl)phenyl]methanol
[4-(Pyrrolidin-1-ylmethyl)phenyl]methanol
APOSOR110012-1G
:
Apollo Scientific
:
[4-(Pyrrolidin-1-ylmethyl)phenyl]methanol
APOSOR110012-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
191.269 g/mol
:
Freezer
:
3267
:
8,III
:
GHS05
:
91271-60-2
:
C₁₂H₁₇NO
:
MFCD08690298