N-Methyl-N-[2-(tetrahydro-2H-pyran-4-yloxy)benzyl]amine
Catalog # APOSOR110010-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:221.295 g/mol
Storage Temperature:Ambient
CAS Number:906352-71-4
Formula:C₁₃H₁₉NO₂
MDL Number:MFCD09064962