0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
3-Ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one
3-Ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one
3-Ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one
APOSOR110002-1G
:
Apollo Scientific
:
3-Ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one
APOSOR110002-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
247.333 g/mol
:
172…176 °C
:
GHS07
:
Ambient
:
71556-74-6
:
C₁₅H₂₁NO₂
:
MFCD00276840