
2-(Aminomethyl)thieno[3,2-b]thiophene hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1172699-35-2
:
Formula:C₇H₇NS₂·ClH
MDL Number:MFCD09702427
:205.728 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together