
7-Aminopyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:68262-33-9
:
Formula:C₇H₆N₄O₂
MDL Number:MFCD00973861
:178.148 g/mol
Melting Pt:242 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together