

1-[4-(4-Nitrophenyl)thiazol-2-yl]thiourea
Catalog # APOSOR10219-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:280.326 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
:GHS08
CAS Number:14015-63-5
Formula:C₁₀H₈N₄O₂S₂
MDL Number:MFCD00245765