Order Entry
Czech Republic
ContactUsLinkComponent
4-Hydroxy-7-methyl-3-phenyl-1H-quinolin-2-one
4-Hydroxy-7-methyl-3-phenyl-1H-quinolin-2-one
Catalog # APOSOR0532-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Hydroxy-7-methyl-3-phenyl-1H-quinolin-2-one
Catalog # APOSOR0532-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :251.28 g/mol
Melting Pt:>300 °C
Storage Temperature:Ambient
CAS Number:83609-87-4
Formula:C₁₆H₁₃NO₂
MDL Number:MFCD00100772