
3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzoic acid
Catalog # APOSOR0508-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:320.383 g/mol
Melting Pt:173…175 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
CAS Number:500013-38-7
Formula:C₁₇H₂₄N₂O₄
MDL Number:MFCD04115219