Order Entry
Czech Republic
ContactUsLinkComponent
N-(1,3-Benzodioxol-5-yl)-N'-(2-chloroethyl)urea
N-(1,3-Benzodioxol-5-yl)-N'-(2-chloroethyl)urea
Catalog # APOSOR01977-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(1,3-Benzodioxol-5-yl)-N'-(2-chloroethyl)urea
Catalog # APOSOR01977-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :242.659 g/mol
Storage Temperature:Ambient
CAS Number:1000018-00-7
Formula:C₁₀H₁₁ClN₂O₃
MDL Number:MFCD09836177