Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
3-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
3-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
APOSOR01975-1G
:
Apollo Scientific
:
3-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
APOSOR01975-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
210.62 g/mol
:
GHS07
:
Ambient
:
1000017-99-1
:
C₈H₇ClN₄O
:
MFCD09842622