
4-(Chloromethyl)-6-hydroxy-2-phenylpyrimidine
Catalog # APOSOR0169-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:220.655 g/mol
Melting Pt:195…198 °C
:GHS07
Storage Temperature:Ambient
CAS Number:35252-98-3
Formula:C₁₁H₉ClN₂O
MDL Number:MFCD00113662