
4-(tert-Butyl)-N,N-dimethylaniline 98%
:2909-79-7
:
:C₁₂H₁₉N
:MFCD00008833
:177.286 g/mol
:250…253 °C
:0.906 g/cm³ (25 °C)
:109 °C
:220-824-6
:GHS07
:Ambient
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals