
2,3-Dihydro-1,4-benzodioxine-2-carboxamidine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85084-36-2
:
Formula:C₉H₁₀N₂O₂·ClH
MDL Number:MFCD00831017
:214.649 g/mol
Melting Pt:180…185 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together