Order Entry
Czech Republic
ContactUsLinkComponent
Palladium 5% Pd on carbon, powder
Irritant
Flammable
Palladium 5% Pd on carbon, powder
Catalog # APOSIN6739-10G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Flammable
Palladium 5% Pd on carbon, powder
Catalog # APOSIN6739-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :106.420 g/mol
Density:2.26 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-115-6
UN:1325
ADR:4.1,III
 :GHS07
 :GHS02
CAS Number:7440-05-3
Formula:Pd
MDL Number:MFCD00011167