m-PEG5-bromide
Catalog # APOSBIPG1606-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:315.201 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₂₃BrO₅
MDL Number:MFCD28950766