Order Entry
Czech Republic
ContactUsLinkComponent
6-(y,y-Dimethylallylamino)purine (2IP) solution 1.0 mg/mL
Irritant
6-(y,y-Dimethylallylamino)purine (2IP) solution 1.0 mg/mL
Catalog # APOSBIPD217-100ML
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-(y,y-Dimethylallylamino)purine (2IP) solution 1.0 mg/mL
Catalog # APOSBIPD217-100ML
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :203.244 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:2365-40-4
Formula:C₁₀H₁₃N₅
MDL Number:MFCD00132998