Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L-(-)-Idose 0.1M aqueous solution
L-(-)-Idose 0.1M aqueous solution
L-(-)-Idose 0.1M aqueous solution
APOSBII593-5MG
:
Apollo Scientific
:
L-(-)-Idose 0.1M aqueous solution
APOSBII593-5MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 mg
Seq:
2210
:
:
180.156 g/mol
:
GHS07
:
Ambient
:
5934-56-5
:
C₆H₁₂O₆
:
MFCD00069825