4-Methylphenyl 2,3,6,2',3',4',6'-hepta-O-acetyl-1-thio-β-D-maltopyranoside ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:27894-92-4
:
Formula:C₃₃H₄₂O₁₇S
MDL Number:MFCD28397805
:742.741 g/mol
Storage Temperature:Ambient
Frequently Bought Together