
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-galactopyranose ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3006-60-8
:
Formula:C₁₆H₂₃NO₁₀
MDL Number:MFCD00214297
:389.355 g/mol
Melting Pt:235…238 °C
:GHS07
Storage Temperature:Freezer
Frequently Bought Together