1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-D-glucopyranose ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:171032-74-9
:
Formula:C₁₄H₁₉N₃O₉
MDL Number:MFCD07778213
:373.315 g/mol
Storage Temperature:Ambient
Frequently Bought Together