
2,4-Dinitrophenyl 2-deoxy-2-fluoro-β-D-glucopyranoside ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:111495-86-4
:
Formula:C₁₂H₁₃FN₂O₉
MDL Number:MFCD00132931
:348.238 g/mol
Melting Pt:83…86 °C
:GHS07
Storage Temperature:Freezer
Frequently Bought Together