Order Entry
Czech Republic
ContactUsLinkComponent
DPDPB (1,2-Di[3'-(2'-pyridyldithio)propionamido]butane)
Irritant
DPDPB (1,2-Di[3'-(2'-pyridyldithio)propionamido]butane)
Catalog # APOSBICL112-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
DPDPB (1,2-Di[3'-(2'-pyridyldithio)propionamido]butane)
Catalog # APOSBICL112-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :482.706 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:141647-62-3
Formula:C₂₀H₂₆N₄O₂S₄
MDL Number:MFCD00153604