
Caerulomycin A
Catalog # APOSBIBR1019-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:229.235 g/mol
Storage Temperature:Freezer
UN:2811
ADR:6.1,III
:GHS06
CAS Number:21802-37-9
Formula:C₁₂H₁₁N₃O₂
MDL Number:MFCD03093715