
4-Aminophenyl β-D-glucuronide, sodium salt
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₁₂H₁₄NO₇·Na
MDL Number:MFCD27975435
:307.232 g/mol
Melting Pt:>180 °C (dec.)
:GHS07
Storage Temperature:Freezer
Frequently Bought Together