ContactUsLinkComponent
PIG® Bio-fluids solidifying absorbent powder
PIG® Bio-fluids solidifying absorbent powder
Catalog # NEWPPLP503
Supplier:  NEW PIG
undefined
PIG® Bio-fluids solidifying absorbent powder
Catalog # NEWPPLP503
Supplier:  NEW PIG
Specifications
  • Description:
    Bio-fluids solidifying absorbent powder
  • Absorbency:
    30,3 L/container
  • Packed:
    4 containers per box
  • Pk:
    4 Bottle
Specifications
About this item
Fast-acting absorbent captures biofluids like blood, urine and vomit and "locks" them inside for cleaner, safer disposal.
  • Permanently solidifies to lock in absorbed liquids so they won't leak or liquefy when the absorbents are handled or placed under pressure
  • Contains chlorine to kill pathogens and reduce odors
  • Clean scent helps reduce offensive odors during and after cleanup
  • Environmentally-friendly product absorbs biological fluids including blood, urine and vomit
  • Forms a solid, non-expanding mass for easier, less-expensive recovery and cleanup
  • Shaker bottle makes it easy to apply absorbent directly onto spill area
High Btu value produces less than 0.1% ash content, ideal for fuels blending and incineration.