Order Entry
Czech Republic
ContactUsLinkComponent
 
Corrosive
Irritant
Platinum(IV) nitrate solution (15% Pt) (w/w)
Catalog # H37737.03
CAS Number:  
Corrosive
Irritant
Platinum(IV) nitrate solution (15% Pt) (w/w)
Catalog # H37737.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :443.10 g/mol
Storage Temperature:Ambient
EINECS:432-400-1
UN:3093
ADR:8,II
 :GHS05
 :GHS07
CAS Number:18496-40-7
Formula:N₄O₁₂Pt
MDL Number:MFCD25372548