ContactUsLinkComponent
Gowns, Tyvek® IsoClean®, model IC702S 00, Non sterile
 :  DuPont
undefined
Gowns, Tyvek® IsoClean®, model IC702S 00, Non sterile
 :  DuPont
  • Catalog No:
  • DUPOIC702SWH00/2XL
  • DUPOIC702SWH00/M
  • Fabric:
  • Tyvek®
  • Tyvek®
  • Closure type:
  • Back fastening
  • Back fastening
  • Gender:
  • Unisex
  • Unisex
  • Seam type:
  • Serged
  • Serged
  • Garment application:
  • Cleanroom, Laboratory
  • Cleanroom, Laboratory
  • Tensile strength:
  • >30 N
  • >30 N
Frequently Bought Together