Benzyl chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:100-44-7
:
Formula:(C₆H₅)CH₂Cl
MDL Number:MFCD00000889
:126.59 g/mol
Storage Temperature:Ambient
UN:1738
ADR:6.1 (8),II
:GHS06
:GHS05
:GHS08
Specification Test Results
- Assay (GC, area%):≥ 99%
- Appearance: Clear colourless to pale yellow liquid:Passes test
- Density (20 °C):1,097 - 1,101
- Acidity:≤ 0,002 meq/g
- Non-volatile matter :≤ 0,05 %
Specification Test Results
Frequently Bought Together