
Citric acid monohydrate for HPLC
Catalog # ACRO464541000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- UV:(1M in water)
- Titration with NaOH:≤99.75 %
Specification Test Results
About this item
:
:210.14 g/mol
Melting Pt:135…152 °C
Flash Pt:174 °C
:GHS07
Storage Temperature:Ambient
CAS Number:5949-29-1
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972