

Tilmicosin phosphate
Catalog # ACRO463270010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- HPLC:≥70 %
Specification Test Results
About this item
:
:967.13 g/mol
Melting Pt:130…135 °C
:GHS08
:GHS07
Storage Temperature:Refrigerator
CAS Number:137330-13-3
Formula:C₄₆H₈₀N₂O₁₃·H₃PO₄
MDL Number:MFCD06797805