Order Entry
Czech Republic
ContactUsLinkComponent
Duloxetine hydrochloride
Health
Duloxetine hydrochloride
Certificates
undefined
Health
Duloxetine hydrochloride

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    162 to 168 °C
  • Loss on drying:
    ≤0.5 % (105 °C, 3 h)
  • HPLC:
    ≥97.5 %
  • Specific optical rotation:
    +116° to +125° (20 °C, 589 nm) (1 % in MeOH)

Specification Test Results

Frequently Bought Together