
Rizatriptan benzoate
Katalogové číslo ACRO462940010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to beige Powder or crystals
- Infrared spectrum:Conforms
- Melting point:178 to 182 °C
- Water:≤0.5 %
- HPLC:≥97.5 %
Specification Test Results
About this item
:
Melting Pt:178…180 °C
:391.475 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:145202-66-0
Formula:C₂₂H₂₅N₅O₂
MDL Number:MFCD00866224