Order Entry
Czech Republic
ContactUsLinkComponent
 
5-Fluoro-5'-deoxyuridine (Doxifluridine)
 
undefined
5-Fluoro-5'-deoxyuridine (Doxifluridine)

 

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    187 to 193 °C
  • Loss on drying:
    ≤0.5 %
  • HPLC:
    ≥97.5 %
  • Specific optical rotation:
    +18° to +22° (20 °C, 589 nm) (1 % in water)

 

Frequently Bought Together