

Nebivolol hydrochloride
Catalog # ACRO462492500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥97.5 %
- HPLC:≥97.5 % (mixture of isomers)
Specification Test Results
About this item
:
:441.90 g/mol
Melting Pt:226 °C
Storage Temperature:Refrigerator
UN:3077
ADR:9,III
:GHS07
:GHS09
CAS Number:152520-56-4
Formula:C₂₂H₂₅F₂NO₄
MDL Number:MFCD07357273