Compact system for 100 ml extraction, KEX 100/KEX 100 F, behrotest®
Dodavatel: BEHR
Certifikáty
O tomto produktu
Complete compact extraction system with basic rack, heating device, holder, tubing and glassware (reaction flasks, extractor, Dimroth condenser for extraction).
- Practical holders for condenser and extraction adapter for secure placement between extractions
- Extractor sizes from 30 to 500 ml
- Condenser with screw connections
- Practical stand for holding condensers securely between extractions
- Holders for secure fixation of extraction adapters after removal of the sample vessels
- S models: The use of extractors with a stopcock does away with the need for any additional distillation after extraction
Infinitely variable heating regulation. After the extraction cycle, the compact system removes the solvent, by means of a discharge valve on the extractor directly into the reservoir. behr equipment for Soxhlet extraction complies with the various requirements of daily laboratory routines.
Frequently Bought Together